Learn More
Sino Biological Human Carboxypeptidase B1 / CPB1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10501H08H20
A DNA sequence encoding the human CPB1 (NP_001862.2) (Met 1-Tyr 417) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5079