Learn More
Sino Biological Human Carboxypeptidase A1 / CPA1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10504H08H10
A DNA sequence encoding the human CPA1 precursor (NP_001859.1) (Met 1-Tyr 419) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5084