Learn More
Sino Biological Human IL17RD Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10507H08H100
A DNA sequence encoding the extracellular domain of human IL17RD (NP_060033.3) precursor (Met 1-Arg 299) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5088