Learn More
Sino Biological Human TREM1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10511H08H50
A DNA sequence encoding the extracellular domain (Met 1-Arg 200) of human TREM1 (NP_061113.1) was fused with the a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5095