Learn More
Sino Biological Human CD140b / PDGFRB Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10514H08H50
A DNA sequence encoding the human PDGFRβ (NP_002600.1) extracellular domain (Met 1-Lys 531) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-5101