Learn More
Sino Biological Human PDGFRB / PDGFR-1 / CD140b Protein DFS Item
Manufacturer:  Sino Biological 10514HCCH200
A DNA sequence encoding the extracellular domain of human PDGFRβ (NP_002600.1) (Met 1-Lys 531) was expressed with six amino acids (ENLYFQ) at the C-terminus.
Catalog No. 50-161-5104