Learn More
Sino Biological Human IL3RA / CD123 Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10518H03H50
A DNA sequence encoding the human IL3RA (NP_002174.1) extracellular domain (Met 1-Arg 305) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5112