Learn More
Sino Biological Human IL3RA / CD123 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10518H08H50
A DNA sequence encoding the human IL3RA (NP_002174.1) extracellular domain (Met 1-Arg 305) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-5114