Learn More
Sino Biological Human SPG21 Protein (GST Tag) DFS Item
Manufacturer:  Sino Biological 10522H09B20
A DNA sequence encoding the full length of human SPG21 (NP_057714.1) (Met 1-Gln 308) was expressed with the GST tag at the N-terminus.
Catalog No. 50-161-5116