Learn More
Sino Biological Human PAPPA2 / Pappalysin 2 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10528H08H50
A DNA sequence encoding the human PAPPA2 mature form (NP_064714.2) corresponding to amino acid (Ser 234-Cys 1396) was expressed, with a carboxy-terminal polyhistidine tag.
Catalog No. 50-161-5124