Learn More
Sino Biological Human MMP1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10532H08H20
A DNA sequence encoding the pro form of human MMP1 enzyme (NP_002412.1) precursor (Met 1-Asn 469) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5132