Learn More
Sino Biological Human PKC iota / PRKCI Protein (GST Tag) DFS Item
Manufacturer:  Sino Biological 10534H09B20
A DNA sequence encoding the human PKCλ (NP_002731.4) (Met 10-Val 596) was fused with the GST tag at the C-terminus.
Catalog No. 50-161-5133