Learn More
Sino Biological Human RAC1 / MIG5 Protein (GST Tag) DFS Item
Manufacturer:  Sino Biological 10535H09B10
A DNA sequence encoding the mature form of human Rac1 isoform A (NP_008839.2) (Met 1-Cys 189) was expressed with the GST tag at the N-terminus.
Catalog No. 50-161-5135