Learn More
Sino Biological Human ACP5 / TRAP Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10550H08H10
A DNA sequence encoding the human ACP5 (NP_001602.1) precursor (Met 1-Pro 320) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5155