Learn More
Sino Biological Human BMPR2 / BMPR-II Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10551H03H100
A DNA sequence encoding the human BMPR-II (NP_001195.2) extracellular domain (Met 1-Ile 151) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5157