Learn More
Sino Biological Human BMPR2 / BMPR-II Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10551H08H50
A DNA sequence encoding the human BMPR-II (NP_001195.2) extracellular domain (Met 1-Ile 151) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5160