Learn More
Sino Biological Human DOPA Decarboxylase / DDC Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10560H08B20
A DNA sequence encoding the full length of human DDC (NP_000781.1) (Met 1-Glu 480) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5182