Learn More
Sino Biological Human TFPI Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10564H08H20
A DNA sequence encoding the human TFPI (NP_006278.1) (Met 1-Lys 282) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5193