Learn More
Sino Biological Human TFPI Protein (His Tag), Biotinylated DFS Item
Manufacturer:  Sino Biological 10564H08HB10
A DNA sequence encoding the human TFPI (NP_006278.1) (Met1-Lys282) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-161-5194