Learn More
Sino Biological Human CD226 / DNAM-1 Protein (His Tag), Biotinylated 20ug
Supplier:  Sino Biological 10565H08HB20
Encompass_Preferred
A DNA sequence encoding the human CD226 (NP_006557.2) (Met1-Asn247) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-161-5200