Learn More
Sino Biological Human TCN2 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10566H08H10
A DNA sequence encoding the human TCN2 (NP_000346.2) (Met 1-Trp 427) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5201