Learn More
Sino Biological Human PTPN2 / TC-PTP Protein (aa 1-314, His & GST Tag) DFS Item
Manufacturer:  Sino Biological 10570H20B50
A DNA sequence encoding the amino acid (Met 1-Asn 314) of human PTPN2 (P17706-1) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-5212