Learn More
Sino Biological Human FGF10 / KGF2 Protein DFS Item
Manufacturer:  Sino Biological 10573HNAE5
A DNA sequence encoding the mature form of human FGF10 (NP_004456.1) (Gln 38-Ser 208) was expressed.
Catalog No. 50-161-5219