Learn More
Sino Biological Human Interleukin-21 / IL-21 Protein DFS Item
Manufacturer:  Sino Biological 10584HNAE20
A DNA sequence encoding the mature form of human IL-21 isoform 1 (Q9HBE4-1) (Gln 23-Ser 155) was expressed and purified, with an initial Met.
Catalog No. 50-161-5239