Learn More
Sino Biological Human CXCL2 / MIP-2 Protein DFS Item
Manufacturer:  Sino Biological 10586HNCE100
A DNA sequence encoding the human CXCL2 (NP_002080.1) (Ala35-Asn107) was expressed.
Catalog No. 50-161-5245