Learn More
Sino Biological Human PCSK9 / NARC1 Protein (His Tag), Biotinylated DFS Item
Manufacturer:  Sino Biological 10594H08HB10
A DNA sequence encoding the human PCSK9 (NP_777596.2, natural mutation Val 474 Ile and Gly 670 Glu) (Met 1-Gln 692) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-161-5255