Learn More
Sino Biological Human PCSK9 / NARC1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10594H08H110
A DNA sequence encoding the human PCSK9 (NP_777596.2, with mutation Asp 374 Tyr, natural mutation Val 474 Ile and Gly 670 Glu) (Met 1-Gln 692) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5252