Learn More
Sino Biological Human LR3 IGF-1 / IGF-I / IGF1 Protein DFS Item
Manufacturer:  Sino Biological 10598HNAY11
A DNA sequence encoding the human IGF1 (P05019, E51R) (Gly49-Ala118) was expressed with a 13 amino acids (MFPAMPLSSLFVN)at its N-terminus.
Catalog No. 50-161-5259