Learn More
Sino Biological Human TNF-alpha / TNFA Protein DFS Item
Manufacturer:  Sino Biological 10602HNAE50
A DNA sequence encoding the human TNF-α soluble form (NP_000585.2) (Val 77-Leu 233) was expressed, with an initial Met at the N-terminus.
Catalog No. 50-161-5268