Learn More
Sino Biological Human Epidermal Growth Factor / EGF Protein DFS Item
Manufacturer:  Sino Biological 10605HNAY1
A DNA sequence encoding the human EGF (NP_001954.2) (Asn971-Arg1023) was expressed.
Catalog No. 50-161-5279