Learn More
Sino Biological Human IL1F6 / IL36A Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10607H07E110
A DNA sequence encoding the mature form of human IL1F6 (Q9UHA7) (Lys 6-Phe158) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-5286