Learn More
Sino Biological Human FGFR1 / CD331 Protein (His & GST Tag) 50ug
Supplier:  Sino Biological 10616H20B150
Encompass_Preferred
A DNA sequence encoding the human FGFR1 isoform 4 (NP_075594.1) cytoplasmic domain (Gly 311-Arg 731) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-5299