Learn More
Sino Biological Human Carbonic Anhydrase XII / CA12 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10617H08H20
A DNA sequence encoding the extracellular domain of human CA12 (NP_001209.1) (Met 1-Gln 291) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5301