Learn More
Sino Biological Human TNFRSF17 / BCMA / CD269 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10620H15H50
A DNA sequence encoding the human TNFRSF17 (NP_001183.2) (Met1-Ala54) was expressed with the Fc region of rabbit IgG at the N-terminus.
Catalog No. 50-161-5310