Thermo Scientific™ 10mm Assembled Clear Autosampler Vial Kits Available on GSA/VA Contract for Federal Government customers only.
Save time and reduce the risk of contamination using this convenience kit. It includes 100 clear vials pre-assembled with 100 screw thread caps and septa.
Manufacturer: Thermo Scientific™ C401067
 View more versions of this product
Catalog No. 03-376-437
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Septa PTFE/Silicone
    Material Glass
    Color Clear
    Patched No
    No. per Pack 100
    Type Assembled Vial Kit
    Vial Closure Size 10-425mm
    Vial Volume (Metric) 2mL
    Closure Color Black
    Item Description Assembled Kit
    Description Clear Glass; Black cap; PTFE/Silicone septum
    Save time during sample preparation and reduce the risk of contamination. Each Thermo Scientific™ National™ 10mm Clear Vial Convenience kits has 100 clear glass vials assembled with 100 screw thread caps and septa. They are packaged in convenient vial trays with clear covers or in economical polybags.
    • Assembled kits include 100 vials with pre-attached caps and septa
    • Packaged in convenient vial trays with clear covers or in polybags
    • Vial trays are stackable for easy storage
    • Vial trays hold vials upright for easy filling and maintaining sample order
  • Description & Specifications
Related Products
Product Content Feedback