Thermo Scientific™ 10mm Autosampler Vial Septa Available on GSA/VA Contract for Federal Government customers only.

Thermo Scientific™ National Target 10mm Septa are designed for use with 10-425 threaded caps.

Manufacturer: Thermo Scientific™ C401055

 View more versions of this product

Catalog No. 03-377-66

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Color White/Blue (Septa)
    Type Pre-Slit,Septum
    Material Silicone/PTFE (Septa)
    Vial Closure Size 10-425mm,10-425mm
    Thickness (Metric) 1.5mm
    Quantity 100
    For Use With Vial Screw Caps
    Item Description Septum for 10-425 Screw Caps

    Thermo Scientific™ National Target 10mm Septa are designed for use with 10-425 threaded caps.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit