Learn More
Thermo Scientific™ 10mm Autosampler Vial Septa Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National Target 10mm Septa are designed for use with 10-425 threaded caps.
Manufacturer:  Thermo Scientific™ C401010
 View more versions of this product
Catalog No. 03-375-12A
  Check Availability
Add to cart
Description
Description
Thermo Scientific™ National Target 10mm Septa are designed for use with 10-425 threaded caps.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
White (Septa)
10-425mm
1000
Vial Screw Caps
Virgin PTFE (Septa)
0.25mm
Septum
Septum for 10-425 Screw Caps
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.