Thermo Scientific™ 10mm Non-Assembled Clear Screw Thread Autosampler Vial Kits Available on GSA/VA Contract for Federal Government customers only.
Save time and reduce the risk of contamination using this convenience kit. Each kit includes 100 clear glass vials and 100 screw thread caps with pre-assembled septa.
Manufacturer: Thermo Scientific™ C401088W
 View more versions of this product
Catalog No. 03-375-15G
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Septa Red PTFE/White Silicone/Red PTFE
    Material Glass
    Color Clear
    Patched Yes
    No. per Pack 100
    Type Unassembled Vial Kit
    Vial Closure Size 10-425mm
    Vial Volume (Metric) 2mL
    Closure Color Light Blue
    Item Description Convenience Kit
    Description Clear Glass; ID patch; Light Blue Cap, Red PTFE/White Silicone/Red PTFE Septum; 2mL
    Thermo Scientific™ National 10mm Clear Vial Convenience kits (Unassembled) save time during sample preparation and reduce the risk of contamination. Each kit includes 100 clear vials and 100 screw thread caps with pre-assembled septa. Reusable two-compartment trays protect vials and closures while keeping matching supplies together. Clear trays to make it easy to keep track of available supplies without opening containers.
    • Two-compartment benchtop storage tray
    • I-D™ vials feature write-on patch with graduations
    • All items remain handy and dust-free
    • Caps with septa are not attached to vials (packed in separate polybag)
    Includes:
    100 vials and 100 caps with pre-assembled septa
  • Description & Specifications
Related Products
Product Content Feedback