Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ 10nm Colloidal Gold Nanoparticles 1OD
10nm colloidal gold nanoparticles ultra high quality, with narrow size distribution and uniform shape
$241.00 - $844.00
Specifications
Content And Storage | Store at 4°C. Do not freeze. |
---|---|
Kit Contents | One 20mL bottle at 1 OD |
Product Type | 10nm Colloidal Gold Nanoparticles 1OD Antibody Labeling Kit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
30-010-020
|
Novus Biologicals™
30010020 |
20 mL |
Each for $241.00
|
|
|||||
30-010-100
|
Novus Biologicals™
30010100 |
100 mL |
Each for $844.00
|
|
|||||
Description
- 10nm colloidal gold nanoparticles are ultra high quality, with narrow size distribution and uniform shape, supplied in range of sizes and concentrations for maximum flexibility
Specifications
Store at 4°C. Do not freeze. | |
One 20mL bottle at 1 OD | |
10nm Colloidal Gold Nanoparticles 1OD Antibody Labeling Kit |
For research use only.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title