Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

11-Chloro-1-undecene 97%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
872-17-3
C11H21Cl
188.739
MFCD00671354
PSEVKFKRYVAODC-UHFFFAOYSA-N
11-chloro-1-undecene,1-undecene, 11-chloro,10-undecenyl chloride,undecylenyl chloride,acmc-1bjnc,undec-10-enyl chloride,11-chloro-1-undecene #,11-chloro-1-undecene
543805
11-chloroundec-1-ene
C=CCCCCCCCCCCl
872-17-3
C11H21Cl
188.739
MFCD00671354
PSEVKFKRYVAODC-UHFFFAOYSA-N
11-chloro-1-undecene,1-undecene, 11-chloro,10-undecenyl chloride,undecylenyl chloride,acmc-1bjnc,undec-10-enyl chloride,11-chloro-1-undecene #,11-chloro-1-undecene
543805
11-chloroundec-1-ene
C=CCCCCCCCCCCl
Quantity 5mL
Infrared Spectrum Authentic
Refractive Index 1.4500 to 1.4520 (20°C, 589nm)
Color Colorless
Packaging Glass bottle
Density 0.9950g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH2CH(CH2)9Cl
Formula Weight 188.74
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 11-Chloro-1-undecene
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.