Learn More
Cayman Chemical 11-ProstaglndIn E1 10mg
SDP
Supplier:  Cayman Chemical 1345010 MG
Encompass_Preferred
11β-PGE1 is an epimerized form of PGE1 at the C-11 position. 11β-PGE1 is a less potent isomer of PGE1. It is 13% and 3.6% as potent as PGE1 in contracting the rat uterus and guinea pig ileum, respectively.{4158}
Catalog No. 50-295-5941