Learn More
Sino Biological Human respiratory syncytial virus (RSV) (A, rsb1734) glycoprotein G / RSV-G Protein (93% Homology) (His Tag) 100ug
Supplier:  Sino Biological 11070V08H2100
Encompass_Preferred
A DNA sequence encoding the glycoprotein G extracellular domain (Asn 66-Arg297) of human respiratory syncytial virus A (93% homologous with strain rsb1734) (P27022-1) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-6460
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.