Learn More
Thermo Scientific™ 11mm Autosampler Vial Crimp Caps, Polyethylene Snap Cap with Silicone/PTFE Seal; Neutral
Minimize contamination from handling with pre-assembled caps and septa; compatible with most HPLC and GC autosamplers.
Manufacturer:  Thermo Scientific™ 11PECST1
Catalog No. 10-800-138
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
Description
Description
Minimize contamination from handling with convenient, pre-assembled Thermo Scientific™ 11mm Autosampler Vial Crimp Caps, compatible with most HPLC and GC autosamplers. Choose from Thermo Scientific, National or Chromacol brands.
Aluminum crimp seals with prefitted septa fit all 11mm crimp top and snap cap vials. Must be applied with a crimping tool.
Ordering Information:
Shipped in sealed polybags to prevent contamination during transport.