Learn More
Thermo Scientific™ 11mm Crimp Top Caps - Routine Range
Use these 11mm Crimp Top Caps as closures for Routine range 11mm Glass Crimp Top Vials in your optimum high throughput HPLC and GC analysis.
$12.12 - $23.23
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
03-251-853
View Documents
Thermo Scientific™
60180ACC11NRT1
11mm crimp cap, silver aluminum, natural rubber/TEF septa, 60 shore, 1.0mm thick, 100/pack Pack of 100 for $12.12
Add to cart
03-251-854
View Documents
Thermo Scientific™
60180ACC11ST1
11mm crimp cap, silver aluminum, silicone/PTFE septa, 60 shore, 1.0mm thick, 100/pack Pack of 100 for $23.23
Add to cart
Description
Description
Use Thermo Scientific™ 11mm Crimp Top Caps as closures for 11mm Glass Crimp Top Vials - Routine range. Use these autosampler vials and closures for optimum performance in standard applications and high throughput analyses. These products exhibit easy handling and high solvent compatibility for the mainstream HPLC and GC applications. Recommended workflows / applications include:
  • Chromatographic applications aligning with all pharmacopeias
  • Lower risk of contamination, degradation and adsorption
  • Standard concentrations
  • Perfect autosampler fit
  • High throughput analysis