Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 11mm Non-Assembled Amber Crimp Top Autosampler Vial Kits Available on GSA/VA Contract for Federal Government customers only.

Save time and reduce the risk of contamination using this convenience kit. Each kit includes 100 amber vials and 100 crimp top caps with pre-assembled septa.

Manufacturer:  Thermo Scientific™ C401288A

 View more versions of this product

Catalog No. 03-375-10B


  Check Availability
Add to cart

Description

Description

Save time during sample preparation and reduce the risk of contamination using Thermo Scientific™ 11mm Amber Crimp Top Vial Convenience Kits (Unassembled). Each kit includes 100 amber glass vials and 100 crimp top caps with pre-assembled septa. Reusable two-compartment trays protect vials and closures while keeping matching supplies together. Clear trays to make it easy to keep track of available supplies without opening containers. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.

  • Include 100 vials (2mL) and separately packed assembled aluminum seals with PTFE/red rubber septa
  • Two-compartment benchtop storage tray
  • All items remain handy and dust free

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Assembled Vial Kit
2mL
Crimp-Top
Amber Glass; Aluminum seal, PTFE/Red Rubber septum; 2mL
Glass
No
11mm
Aluminum
Convenience Kit, Standard Opening Crimp Top Vial
PTFE/Red Rubber
Amber
500
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.