Learn More
CD365 (TIM1) Mouse anti-Human, PE, Clone: 1D12-TIM1, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
 View more versions of this product
Catalog No. 12-365-941
Add to cart
Description
Description
Mouse Monoclonal Antibody
CD365 (TIM1) Monoclonal antibody specifically detects CD365 (TIM1) in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
CD365 (TIM1)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q96D42
AI503787; CD365; HAVCR; Havcr1; HAVCR-1; Hepatitis A virus cellular receptor 1; Hepatitis A virus cellular receptor 1 homolog; Kidney injury molecule 1; KIM1; KIM-1; T cell immunoglobulin and mucin domain-containing protein 1; T cell membrane protein 1; T-cell immunoglobulin and mucin domain-containing protein 1; T-cell immunoglobulin mucin receptor 1; T-cell membrane protein 1; TIM; TIM1; TIM-1; Timd1; TIMD-1
Mouse
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
26762
Flow Cytometry
1D12-TIM1
PE
Havcr1
Liquid
Havcr1
IgV domain of human CD365
Affinity chromatography
RUO
Antibody
Monoclonal
Human