Learn More
TCR V beta 11 Rat anti-Mouse, PE, Clone: RR3-15, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
 View more versions of this product
Catalog No. 12-582-782
Add to cart
Description
Description
Rat Monoclonal Antibody
TCR V beta 11 Monoclonal antibody specifically detects TCR V beta 11 in Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
TCR V beta 11
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
TCR V beta11; TCRV beta 11; TCRV beta11; Vb11; Vbeta11
IgG2b, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
Mouse
Flow Cytometry
RR3-15
PE
Liquid
Rat
Affinity chromatography
RUO
Antibody
Monoclonal