Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 12-Aminododecanoic acid, 96%

Manufacturer:  Alfa Aesar™ B2512114

Quantity 25g

 View more versions of this product

Catalog No. AAB2512114


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

693-57-2
215.34
12-Aminolauric acid
H2N(CH2)11CO2H
MFCD00008153
12-Aminododecanoic acid

Specifications

693-57-2
25g
215.34
12-Aminolauric acid
96%
185° to 187°C
H2N(CH2)11CO2H
MFCD00008153
12-Aminododecanoic acid

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit