Alfa Aesar™ 12-Aminododecanoic acid, 96%

Manufacturer: Alfa Aesar™ B2512103

Quantity 1g

 View more versions of this product

Catalog No. AAB2512103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 693-57-2
    Molecular Formula H2N(CH2)11CO2H
    Formula Weight 215.34
    MDL Number MFCD00008153
    Synonym 12-Aminolauric acid
    Chemical Name or Material 12-Aminododecanoic acid

    Specifications

    Formula Weight 215.34
    Melting Point 185° to 187°C
    Quantity 1g
    TSCA TSCA
    Molecular Formula H2N(CH2)11CO2H
    MDL Number MFCD00008153
    CAS 693-57-2
    Synonym 12-Aminolauric acid
    Chemical Name or Material 12-Aminododecanoic acid
    Assay 96%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit