Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 12-Aminododecanoic acid, 96%

Click to view available options
Quantity:
1g
5g
25g
693-57-2
C12H25NO2
215.337
MFCD00008153
PBLZLIFKVPJDCO-UHFFFAOYSA-N
12-aminolauric acid,12-amino-dodecanoic acid,dodecanoic acid, 12-amino,ccris 6171,omega-aminododecanoic acid,12-aminolauricacid,omega-aminolauric acid,12-aminododecanic acid,.omega.-aminolauric acid
69661
12-aminododecanoic acid
C(CCCCCC(=O)O)CCCCCN
693-57-2
C12H25NO2
215.337
MFCD00008153
PBLZLIFKVPJDCO-UHFFFAOYSA-N
12-aminolauric acid,12-amino-dodecanoic acid,dodecanoic acid, 12-amino,ccris 6171,omega-aminododecanoic acid,12-aminolauricacid,omega-aminolauric acid,12-aminododecanic acid,.omega.-aminolauric acid
69661
12-aminododecanoic acid
C(CCCCCC(=O)O)CCCCCN
Quantity 5g
Melting Point 185°C to 187°C
Formula Weight 215.34
Percent Purity 96%
Chemical Name or Material 12-Aminododecanoic acid